4-Methyl-1,5,6,7-tetrahydro-2H-cyclopenta[b]pyridin-2-one ≥95%

Supplier: Matrix Scientific

054810-500MG 054810-1G
101908-624EA 278.46 USD
101908-624 101908-626
4-Methyl-1,5,6,7-tetrahydro-2H-cyclopenta[b]pyridin-2-one ≥95%
4-Methyl-1,5,6,7-tetrahydro-2H-cyclopenta[b]pyridin-2-one

Hazard: Irritant

Formula: C₉H₁₁NO
MW: 149.19 g/mol
Melting Pt: 244…246 °C
MDL Number: MFCD16140309
CAS Number: 20594-30-3

Order Now

Specification Test Results

Melting point 244-246°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR