Methyl-2-{3-[(tert-butoxycarbonyl)amino]piperidino}-5-nitrobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

054808-500MG 054808-1G
101908-614EA 375.38 USD
101908-614 101908-616
Methyl-2-{3-[(tert-butoxycarbonyl)amino]piperidino}-5-nitrobenzenecarboxylate ≥95%
Methyl-2-{3-[(tert-butoxycarbonyl)amino]piperidino}-5-nitrobenzenecarboxylate

Hazard: Irritant

Formula: C₁₈H₂₅N₃O₆
MW: 379.42 g/mol
Melting Pt: 107…108 °C
MDL Number: MFCD16140307
CAS Number: 1221791-90-7

Order Now

Specification Test Results

Melting point 107-108°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR