Methyl-4-methyl-2-(methylsulfonyl)-5-pyrimidinecarboxylate ≥95%
Supplier: Matrix Scientific
054798-500MG
054798-1G
101908-574EA
529.23
USD
101908-574
101908-576
Methyl-4-methyl-2-(methylsulfonyl)-5-pyrimidinecarboxylate ≥95%
Methyl-4-methyl-2-(methylsulfonyl)-5-pyrimidinecarboxylate
Hazard: Irritant
Formula:
C₈H₁₀N₂O₄S MW: 230.24 g/mol Melting Pt: 99…100 °C |
MDL Number:
MFCD16140304 CAS Number: 1221792-75-1 |
Specification Test Results
Melting point | 99-100°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...