Methyl-2-hydroxy-5-methoxy-3-nitrobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-hydroxy-5-methoxy-3-nitrobenzoate

054795-500MG 054795-1G 054795-5G
101908-560EA 278.46 USD
101908-560 101908-562 101908-564
Methyl-2-hydroxy-5-methoxy-3-nitrobenzenecarboxylate ≥95%
Methyl-2-hydroxy-5-methoxy-3-nitrobenzenecarboxylate

Hazard: Irritant

Formula: C₉H₉NO₆
MW: 227.18 g/mol
Melting Pt: 139…141 °C
MDL Number: MFCD16140302
CAS Number: 2888-09-7

Order Now

Specification Test Results

Melting point 139-141°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR