5-(3-Methyl-2,5-dioxo-1-imidazolidinyl)pentanohydrazide ≥95%

Supplier: Matrix Scientific

054794-500MG 054794-1G
101908-556EA 375.38 USD
101908-556 101908-558
5-(3-Methyl-2,5-dioxo-1-imidazolidinyl)pentanohydrazide ≥95%
5-(3-Methyl-2,5-dioxo-1-imidazolidinyl)pentanohydrazide

Hazard: Irritant

Formula: C₉H₁₆N₄O₃
MW: 228.25 g/mol
Melting Pt: 73…75 °C
MDL Number: MFCD16140301
CAS Number: 1221792-87-5

Order Now

Specification Test Results

Melting point 73-75°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR