Methyl-2,5-dimethyl-1,3-benzoxazole-7-carboxylate ≥95%

Supplier: Matrix Scientific

054793-500MG 054793-1G 054793-5G
101908-550EA 556.92 USD
101908-550 101908-552 101908-554
Methyl-2,5-dimethyl-1,3-benzoxazole-7-carboxylate ≥95%
Methyl-2,5-dimethyl-1,3-benzoxazole-7-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₁NO₃
MW: 205.22 g/mol
Melting Pt: 110…112 °C
MDL Number: MFCD16140300
CAS Number: 1221792-61-5

Order Now

Specification Test Results

Melting point 110-112°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR