Methyl-2-(chloromethyl)-7-methoxy-1,3-benzoxazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

054792-500MG 054792-1G
101908-546EA 747.69 USD
101908-546 101908-548
Methyl-2-(chloromethyl)-7-methoxy-1,3-benzoxazole-5-carboxylate ≥95%
Methyl-2-(chloromethyl)-7-methoxy-1,3-benzoxazole-5-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₀ClNO₄
Melting Pt: 114…116 °C
MDL Number: MFCD16140299
CAS Number: 1221792-62-6

Order Now

Specification Test Results

Melting point 114-116°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR