Methyl-2-(chloromethyl)-5-methoxy-1,3-benzoxazole-7-carboxylate ≥95%

Supplier: Matrix Scientific

054791-500MG 054791-1G
101908-542EA 921.54 USD
101908-542 101908-544
Methyl-2-(chloromethyl)-5-methoxy-1,3-benzoxazole-7-carboxylate ≥95%
Methyl-2-(chloromethyl)-5-methoxy-1,3-benzoxazole-7-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₀ClNO₄
Melting Pt: 128…130 °C
MDL Number: MFCD16140298
CAS Number: 1221792-89-7

Order Now

Specification Test Results

Melting point 128-130°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR