Methyl-2-(chloromethyl)-5-methyl-1,3-benzoxazole-7-carboxylate ≥95%

Supplier: Matrix Scientific

054790-500MG 054790-1G
101908-538EA 556.92 USD
101908-538 101908-540
Methyl-2-(chloromethyl)-5-methyl-1,3-benzoxazole-7-carboxylate ≥95%
Methyl-2-(chloromethyl)-5-methyl-1,3-benzoxazole-7-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₀ClNO₃
Melting Pt: 81…83 °C
MDL Number: MFCD16140297
CAS Number: 1221792-26-2

Order Now

Specification Test Results

Melting point 81-83°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR