Methyl-5-bromo-2-methyl-1,3-benzoxazole-7-carboxylate ≥95%

Supplier: Matrix Scientific

054788-500MG 054788-1G
101908-530EA 546.15 USD
101908-530 101908-532
Methyl-5-bromo-2-methyl-1,3-benzoxazole-7-carboxylate ≥95%
Methyl-5-bromo-2-methyl-1,3-benzoxazole-7-carboxylate

Hazard: Irritant

Formula: C₁₀H₈BrNO₃
Melting Pt: 141…143 °C
MDL Number: MFCD16140295
CAS Number: 1221792-65-9

Order Now

Specification Test Results

Melting point 141-143°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR