Methyl-5-bromo-2-(chloromethyl)-1,3-benzoxazole-7-carboxylate ≥95%
Supplier: Matrix Scientific
054787-500MG
054787-1G
101908-526EA
1109.23
USD
101908-526
101908-528
Methyl-5-bromo-2-(chloromethyl)-1,3-benzoxazole-7-carboxylate ≥95%
Methyl-5-bromo-2-(chloromethyl)-1,3-benzoxazole-7-carboxylate
Hazard: Irritant
Formula:
C₁₀H₇BrClNO₃ MW: 304.53 g/mol Melting Pt: 113…115 °C |
MDL Number:
MFCD16140294 CAS Number: 1221792-54-6 |
Specification Test Results
Melting point | 113-115°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...