Methyl-4-amino-5-iodo-2-methoxybenzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-amino-5-iodo-2-methoxybenzoate

054785-500MG 054785-1G 054785-5G
101908-518EA 356.92 USD
101908-518 101908-520 101908-522
Methyl-4-amino-5-iodo-2-methoxybenzenecarboxylate ≥95%
Methyl-4-amino-5-iodo-2-methoxybenzenecarboxylate

Hazard: Irritant

Formula: C₉H₁₀INO₃
MW: 307.09 g/mol
Melting Pt: 160…164 °C
MDL Number: MFCD11112895
CAS Number: 137832-56-5

Order Now

Specification Test Results

Melting point 160-164°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR