2-(Isopropylamino)-5-nitrobenzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

054779-500MG 054779-1G
101908-496EA 556.92 USD
101908-496 101908-498
2-(Isopropylamino)-5-nitrobenzenecarboxylic acid ≥95%
2-(Isopropylamino)-5-nitrobenzenecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₄
Melting Pt: 229…230 °C
MDL Number: MFCD11131983
CAS Number: 1019387-83-7

Order Now

Specification Test Results

Melting point 229-230°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR