Ethyl-2-fluoro-5-nitrobenzenecarboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-fluoro-5-nitrobenzoate

054740-500MG 054740-1G 054740-5G
101908-342EA 107.69 USD
101908-342 101908-344 101908-346
Ethyl-2-fluoro-5-nitrobenzenecarboxylate ≥97%
Ethyl-2-fluoro-5-nitrobenzenecarboxylate

Hazard: Irritant

Formula: C₉H₈FNO₄
MW: 213.17 g/mol
Melting Pt: 53…55 °C
MDL Number: MFCD06204381
CAS Number: 367-79-3

Order Now

Specification Test Results

Melting point 53-55°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR