3,4-Dihydro-1H-spiro[naphthalene-2,2'-[1,3]thiazolidine]-4'-one ≥95%

Supplier: Matrix Scientific

054717-500MG 054717-1G
101908-250EA 278.46 USD
101908-250 101908-252
3,4-Dihydro-1H-spiro[naphthalene-2,2'-[1,3]thiazolidine]-4'-one ≥95%
3,4-Dihydro-1H-spiro[naphthalene-2,2'-[1,3]thiazolidine]-4'-one

Hazard: Irritant

Formula: C₁₂H₁₃NOS
Melting Pt: 200…201 °C
MDL Number: MFCD16140281
CAS Number: 1221792-37-5

Order Now

Specification Test Results

Melting point 200-201°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR