3',4'-Dihydro-1'H,3H-spiro[1,3-benzothiazole-2,2'-naphthalene] ≥95%

Supplier: Matrix Scientific

054716-500MG 054716-1G
101908-246EA 278.46 USD
101908-246 101908-248
3',4'-Dihydro-1'H,3H-spiro[1,3-benzothiazole-2,2'-naphthalene] ≥95%
3',4'-Dihydro-1'H,3H-spiro[1,3-benzothiazole-2,2'-naphthalene]

Hazard: Irritant

Formula: C₁₆H₁₅NS
Melting Pt: 121…122 °C
MDL Number: MFCD16140280
CAS Number: 1221792-12-6

Order Now

Specification Test Results

Melting point 121-122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR