2,5-Dichloronicotinamide ≥95%

Supplier: Matrix Scientific

054708-500MG 054708-1G 054708-5G
101908-218EA 181.54 USD
101908-218 101908-220 101908-222
2,5-Dichloronicotinamide ≥95%
2,5-Dichloronicotinamide

Hazard: Irritant

Formula: C₆H₄Cl₂N₂O
Melting Pt: 171-173°C
MDL Number: MFCD00204159
CAS Number: 75291-86-0

Order Now

Specification Test Results

Melting point 171-173°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR