3,8-Dibromoimidazo[1,2-a]pyridine-6-carbonitrile ≥95%

Supplier: Matrix Scientific

054707-500MG 054707-1G
101908-214EA 921.54 USD
101908-214 101908-216
3,8-Dibromoimidazo[1,2-a]pyridine-6-carbonitrile ≥95%
3,8-Dibromoimidazo[1,2-a]pyridine-6-carbonitrile

Hazard: Irritant

Formula: C₈H₃Br₂N₃
Melting Pt: 195…198 °C
MDL Number: MFCD16140277
CAS Number: 1221792-63-7

Order Now

Specification Test Results

Melting point 195-198°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR