5-(Cyanomethyl)-2-pyridine carboxylic acid ≥95%
Supplier: Matrix Scientific
054706-500MG
054706-1G
101908-210EA
404.62
USD
101908-210
101908-212
5-(Cyanomethyl)-2-pyridine carboxylic acid ≥95%
5-(Cyanomethyl)-2-pyridine carboxylic acid
Hazard: Irritant
Formula:
C₈H₆N₂O₂ Melting Pt: 225 °C (dec.) |
MDL Number:
MFCD16140276 CAS Number: 1221791-92-9 |
Specification Test Results
Melting point | 225°Cdec |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...