2-Chloro-1-(1H-pyrrolo[2,3-b]pyridin-3-yl)-1-ethanone ≥97%

Supplier: Matrix Scientific

054705-500MG 054705-1G
101908-206EA 556.92 USD
101908-206 101908-208
2-Chloro-1-(1H-pyrrolo[2,3-b]pyridin-3-yl)-1-ethanone ≥97%
2-Chloro-1-(1H-pyrrolo[2,3-b]pyridin-3-yl)-1-ethanone

Hazard: Irritant

Formula: C₉H₇ClN₂O
Melting Pt: 300 °C (dec.)
MDL Number: MFCD09743458
CAS Number: 83393-47-9

Order Now

Specification Test Results

Melting point 300°C(Dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR