5-Chloro-2-methoxy-3-nitrophenylbenzenecarboxylate ≥95%

Supplier: Matrix Scientific

054701-500MG 054701-1G
101908-192EA 921.54 USD
101908-192 101908-194
5-Chloro-2-methoxy-3-nitrophenylbenzenecarboxylate ≥95%
5-Chloro-2-methoxy-3-nitrophenylbenzenecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₀ClNO₅
Melting Pt: 123…125 °C
MDL Number: MFCD16140275
CAS Number: 1221792-84-2

Order Now

Specification Test Results

Melting point 123-125°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR