5-Bromo-2,2-dimethyl-1,3-dioxane-4,6-dione (5-Bromomeldrum's acid) ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Bromo-2,2-dimethyl-1,3-dioxane-4,6-dione
054691-500MG
054691-1G
054691-5G
101908-148EA
200
USD
101908-148
101908-150
101908-152
5-Bromo-2,2-dimethyl-1,3-dioxane-4,6-dione (5-Bromomeldrum's acid) ≥95%
5-Bromo-2,2-dimethyl-1,3-dioxane-4,6-dione (5-Bromomeldrum's acid)
Hazard: Irritant
Formula:
C₆H₇BrO₄ Melting Pt: 84-85°C |
CAS Number:
66145-20-8 |
Specification Test Results
Melting point | 84-85°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...