5-Bromo-3-cyano-4,6-dimethyl-2-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

054690-500MG 054690-1G
101908-144EA 1109.23 USD
101908-144 101908-146
5-Bromo-3-cyano-4,6-dimethyl-2-pyridine carboxylic acid ≥95%
5-Bromo-3-cyano-4,6-dimethyl-2-pyridine carboxylic acid

Hazard: Irritant

Formula: C₉H₇BrN₂O₂
MW: 255.08 g/mol
Melting Pt: 187 °C (dec.)
MDL Number: MFCD16140267
CAS Number: 1221792-64-8

Order Now

Specification Test Results

Melting point 187°CDec

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR