8-Benzyl-1-thia-4,8-diazaspiro[4.5]decan-3-one ≥95%

Supplier: Matrix Scientific

054685-500MG 054685-1G
101908-124EA 278.46 USD
101908-124 101908-126
8-Benzyl-1-thia-4,8-diazaspiro[4.5]decan-3-one ≥95%
8-Benzyl-1-thia-4,8-diazaspiro[4.5]decan-3-one

Hazard: Irritant

Formula: C₁₄H₁₈N₂OS
Melting Pt: 168…170 °C
MDL Number: MFCD16140266
CAS Number: 32533-11-2

Order Now

Specification Test Results

Melting point 168-170°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR