2-(Benzyloxy)-5-chloro-3-nitrophenylbenzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Benzyloxy)-5-chloro-3-nitrophenyl benzoate

054682-500MG 054682-1G
101908-112EA 747.69 USD
101908-112 101908-114
2-(Benzyloxy)-5-chloro-3-nitrophenylbenzenecarboxylate ≥95%
2-(Benzyloxy)-5-chloro-3-nitrophenylbenzenecarboxylate

Hazard: Irritant

Formula: C₂₀H₁₄ClNO₅
MW: 383.79 g/mol
Melting Pt: 134…136 °C
MDL Number: MFCD16140265
CAS Number: 891497-54-4

Order Now

Specification Test Results

Melting point 134-136°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR