3-(Benzyloxy)-5-bromo-2-methyl-4-pyridinol ≥95%

Supplier: Matrix Scientific

054681-500MG 054681-1G 054681-5G
101908-106EA 556.92 USD
101908-106 101908-108 101908-110
3-(Benzyloxy)-5-bromo-2-methyl-4-pyridinol ≥95%
3-(Benzyloxy)-5-bromo-2-methyl-4-pyridinol

Hazard: Irritant

Formula: C₁₃H₁₂BrNO₂
Melting Pt: 197…199 °C
MDL Number: MFCD16140264
CAS Number: 906657-86-1

Order Now

Specification Test Results

Melting point 197-199°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR