2-Amino-5-(trifluoromethyl)-3-[2-(trimethylsilyl)ethynyl]benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

054678-500MG 054678-1G
101908-094EA 747.69 USD
101908-094 101908-096
2-Amino-5-(trifluoromethyl)-3-[2-(trimethylsilyl)ethynyl]benzenecarbonitrile ≥95%
2-Amino-5-(trifluoromethyl)-3-[2-(trimethylsilyl)ethynyl]benzenecarbonitrile

Hazard: Irritant

Formula: C₁₃H₁₃F₃N₂Si
Melting Pt: 113…114 °C
MDL Number: MFCD16140263
CAS Number: 1221792-67-1

Order Now

Specification Test Results

Melting point 113-114°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR