1-(4-Amino-3-methoxyphenyl)-1-ethanol ≥95%

Supplier: Matrix Scientific

054674-500MG 054674-1G
101908-080EA 747.69 USD
101908-080 101908-082
1-(4-Amino-3-methoxyphenyl)-1-ethanol ≥95%
1-(4-Amino-3-methoxyphenyl)-1-ethanol

Hazard: Irritant

Formula: C₉H₁₃NO₂
Melting Pt: 67…69 °C
MDL Number: MFCD16140262
CAS Number: 1221792-01-3

Order Now

Specification Test Results

Melting point 67-69°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR