5-Acetyl-2-pyridine carboxylic acid ≥97%

Supplier: Matrix Scientific

054664-500MG 054664-1G
101908-042EA 600 USD
101908-042 101908-044
5-Acetyl-2-pyridine carboxylic acid ≥97%
5-Acetyl-2-pyridine carboxylic acid

Hazard: Irritant

Formula: C₈H₇NO₃
MW: 165.15 g/mol
Melting Pt: 179 °C (dec.)
MDL Number: MFCD10697704
CAS Number: 65184-39-6

Order Now

Specification Test Results

Melting point 179°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR