3-Bromo-2-hydroxy-5-nitropyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-5-nitro-2(1H)-pyridinone, 3-Bromo-5-nitro-2-pyridinol, 3-Bromo-5-nitro-2-pyridone

054582-1G 054582-5G
101907-852EA 396.92 USD
101907-852 101907-854
3-Bromo-2-hydroxy-5-nitropyridine ≥98%
3-Bromo-2-hydroxy-5-nitropyridine

Hazard: Irritant/Refrigerate

Formula: C5H3BrN2O3
MW: 218.99 g/mol
Melting Pt: 214…218 °C
MDL Number: MFCD03840431
CAS Number: 15862-33-6

Order Now

Specification Test Results

Melting point 214-218°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR