3-Bromo-6-methyl-5-nitro-1H-indazole ≥95%
Supplier: Matrix Scientific
054581-500MG
054581-1G
101907-848EA
747.69
USD
101907-848
101907-850
3-Bromo-6-methyl-5-nitro-1H-indazole ≥95%
3-Bromo-6-methyl-5-nitro-1H-indazole
Hazard: Irritant
Formula:
C₈H₆BrN₃O₂ Melting Pt: 223…225 °C |
MDL Number:
MFCD09026989 CAS Number: 1000343-58-7 |
Specification Test Results
Melting point | 223-225°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...