[2-(Chloromethyl)-1,3-benzoxazol-5-yl]methyl acetate ≥95%

Supplier: Matrix Scientific

054577-500MG 054577-1G 054577-5G
101907-830EA 747.69 USD
101907-830 101907-832 101907-834
[2-(Chloromethyl)-1,3-benzoxazol-5-yl]methyl acetate ≥95%
[2-(Chloromethyl)-1,3-benzoxazol-5-yl]methyl acetate

Hazard: Irritant

Formula: C₁₁H₁₀ClNO₃
Melting Pt: 70…72 °C
MDL Number: MFCD14584775
CAS Number: 1221791-61-2

Order Now

Specification Test Results

Melting point 70-72°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR