3-Bromo-6-chloro-8-iodoimidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

054575-500MG 054575-1G
101907-822EA 921.54 USD
101907-822 101907-824
3-Bromo-6-chloro-8-iodoimidazo[1,2-a]pyridine ≥95%
3-Bromo-6-chloro-8-iodoimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₇H₃BrClIN₂
Melting Pt: 144-145°C
MDL Number: MFCD14584773
CAS Number: 1221791-77-0

Order Now

Specification Test Results

Melting point 144-145°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR