2-(Ethylamino)-6-fluorobenzenecarbonitrile ≥95%

Supplier: Matrix Scientific

054570-500MG 054570-1G
101907-808EA 278.46 USD
101907-808 101907-810
2-(Ethylamino)-6-fluorobenzenecarbonitrile ≥95%
2-(Ethylamino)-6-fluorobenzenecarbonitrile

Hazard: Irritant

Formula: C₉H₉FN₂
Melting Pt: 47…49 °C
MDL Number: MFCD12193244
CAS Number: 119584-72-4

Order Now

Specification Test Results

Melting point 47-49°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR