N-(2-Aminobenzyl)-2-nitroaniline ≥95%

Supplier: Matrix Scientific

054569-500MG 054569-1G
101907-804EA 278.46 USD
101907-804 101907-806
N-(2-Aminobenzyl)-2-nitroaniline ≥95%
N-(2-Aminobenzyl)-2-nitroaniline

Hazard: Irritant

Formula: C₁₃H₁₃N₃O₂
Melting Pt: 101…103 °C
MDL Number: MFCD14584851
CAS Number: 1221792-14-8

Order Now

Specification Test Results

Melting point 101-103°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR