N-(4-Aminobenzyl)-2-nitroaniline ≥95%

Supplier: Matrix Scientific

054568-500MG 054568-1G
101907-800EA 278.46 USD
101907-800 101907-802
N-(4-Aminobenzyl)-2-nitroaniline ≥95%
N-(4-Aminobenzyl)-2-nitroaniline

Hazard: Irritant

Formula: C₁₃H₁₃N₃O₂
Melting Pt: 120…122 °C
MDL Number: MFCD14584850
CAS Number: 1221792-11-5

Order Now

Specification Test Results

Melting point 120-122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR