tert-Butyl-4-(3-cyano-6-isopropyl-2-pyridinyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%

Supplier: Matrix Scientific

054567-500MG 054567-1G 054567-5G
101907-794EA 747.69 USD
101907-794 101907-796 101907-798
tert-Butyl-4-(3-cyano-6-isopropyl-2-pyridinyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
tert-Butyl-4-(3-cyano-6-isopropyl-2-pyridinyl)tetrahydro-1(2H)-pyrazinecarboxylate

Hazard: Irritant

Formula: C₁₈H₂₆N₄O₂
MW: 330.43 g/mol
Melting Pt: 74…75 °C
MDL Number: MFCD14584849
CAS Number: 1221792-09-1

Order Now

Specification Test Results

Melting point 74-75°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR