Methyl-3-amino-5-bromo-2-methoxybenzoate hydrochloride ≥95%

Supplier: Matrix Scientific

054562-500MG 054562-1G
101907-774EA 747.69 USD
101907-774 101907-776
Methyl-3-amino-5-bromo-2-methoxybenzoate hydrochloride ≥95%
Methyl-3-amino-5-bromo-2-methoxybenzoate hydrochloride

Hazard: Irritant

Formula: C₉H₁₁BrClNO₃
MW: 296.55 g/mol
Melting Pt: 199…201 °C
MDL Number: MFCD14584846
CAS Number: 926036-31-9

Order Now

Specification Test Results

Melting point 199-201°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR