6-(2-Methyl-1,3-dioxolan-2-yl)-2-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

054559-500MG 054559-1G
101907-764EA 341.54 USD
101907-764 101907-766
6-(2-Methyl-1,3-dioxolan-2-yl)-2-pyridine carboxylic acid ≥95%
6-(2-Methyl-1,3-dioxolan-2-yl)-2-pyridine carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₁NO₄
MW: 209.2 g/mol
Melting Pt: 113…115 °C
MDL Number: MFCD14584843
CAS Number: 122637-38-1

Order Now

Specification Test Results

Melting point 113-115°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR