1,2,6,7-Tetrahydro-8H-indeno[5,4-b]furan-8-one ≥95%

Supplier: Matrix Scientific

Synonyms: 6,7-Dihydro-1H-indeno[5,4-b]furan-8(2H)-one, Ramelteon Impurity 5, 1,2,6,7-Tetrahydro-8H-indeno[5,4-b]furan-8-one

054557-500MG 054557-1G
101907-756EA 1109.23 USD
101907-756 101907-758
1,2,6,7-Tetrahydro-8H-indeno[5,4-b]furan-8-one ≥95%
1,2,6,7-Tetrahydro-8H-indeno[5,4-b]furan-8-one

Hazard: Irritant

Formula: C₁₁H₁₀O₂
MW: 174.2 g/mol
Melting Pt: 133…135 °C
MDL Number: MFCD09955085
CAS Number: 196597-78-1

Order Now

Specification Test Results

Melting point 133-135°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR