8-Methyl-1-thia-4,8-diazaspiro[4.5]decane hydrochloride ≥95%

Supplier: Matrix Scientific

054547-500MG 054547-1G 054547-5G
101907-722EA 278.46 USD
101907-722 101907-724 101907-726
8-Methyl-1-thia-4,8-diazaspiro[4.5]decane hydrochloride ≥95%
8-Methyl-1-thia-4,8-diazaspiro[4.5]decane hydrochloride

Hazard: Irritant

Formula: C₈H₁₇ClN₂S
Melting Pt: 189…191 °C
MDL Number: MFCD14584835
CAS Number: 1221575-00-3

Order Now

Specification Test Results

Melting point 189-191°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR