5-Bromo-2-methoxy-4,6-dimethylnicotinonitrile ≥95%

Supplier: Matrix Scientific

054546-500MG 054546-1G
101907-718EA 556.92 USD
101907-718 101907-720
5-Bromo-2-methoxy-4,6-dimethylnicotinonitrile ≥95%
5-Bromo-2-methoxy-4,6-dimethylnicotinonitrile

Hazard: Irritant

Formula: C₉H₉BrN₂O
Melting Pt: 103…105 °C
MDL Number: MFCD03787138
CAS Number: 113893-02-0

Order Now

Specification Test Results

Melting point 103-105°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR