2-Bromo-1-(3-methoxy-4-nitrophenyl)-1-ethanone ≥90%
Supplier: Matrix Scientific
054545-500MG
054545-1G
101907-714EA
307.69
USD
101907-714
101907-716
2-Bromo-1-(3-methoxy-4-nitrophenyl)-1-ethanone ≥90%
2-Bromo-1-(3-methoxy-4-nitrophenyl)-1-ethanone
Hazard: Irritant
Formula:
C₉H₈BrNO₄ Melting Pt: 98…100 °C |
MDL Number:
MFCD14584834 CAS Number: 90725-63-6 |
Specification Test Results
Melting point | 98-100°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...