1-(4-Amino-3-bromophenyl)-2,2,2-trifluoro-1-ethanone ≥95%

Supplier: Matrix Scientific

054541-500MG 054541-1G
101907-698EA 480 USD
101907-698 101907-700
1-(4-Amino-3-bromophenyl)-2,2,2-trifluoro-1-ethanone ≥95%
1-(4-Amino-3-bromophenyl)-2,2,2-trifluoro-1-ethanone

Hazard: Irritant

Formula: C₈H₅BrF₃NO
Melting Pt: 130…132 °C
MDL Number: MFCD14584831
CAS Number: 1221572-25-3

Order Now

Specification Test Results

Melting point 130-132°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR