Ethyl-5-bromo-3-cyano-4,6-dimethyl-2-pyridinecarboxylate ≥95%
Supplier: Matrix Scientific
054540-500MG
054540-1G
101907-694EA
747.69
USD
101907-694
101907-696
Ethyl-5-bromo-3-cyano-4,6-dimethyl-2-pyridinecarboxylate ≥95%
Ethyl-5-bromo-3-cyano-4,6-dimethyl-2-pyridinecarboxylate
Hazard: Irritant
Formula:
C₁₁H₁₁BrN₂O₂ Melting Pt: 84…86 °C |
MDL Number:
MFCD14584830 CAS Number: 1221791-96-3 |
Specification Test Results
Melting point | 84-86°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...