3-Bromo-8-chloroimidazo[1,2-a]pyridine-6-carbonitrile ≥95%

Supplier: Matrix Scientific

054537-500MG 054537-1G
101907-680EA 921.54 USD
101907-680 101907-682
3-Bromo-8-chloroimidazo[1,2-a]pyridine-6-carbonitrile ≥95%
3-Bromo-8-chloroimidazo[1,2-a]pyridine-6-carbonitrile

Hazard: Irritant

Formula: C₈H₃BrClN₃
Melting Pt: 179-181°C
MDL Number: MFCD14584827
CAS Number: 1221791-70-3

Order Now

Specification Test Results

Melting point 179-181°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR