3-Cyano-4,6-dimethyl-2-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

054536-500MG 054536-1G
101907-676EA 556.92 USD
101907-676 101907-678
3-Cyano-4,6-dimethyl-2-pyridine carboxylic acid ≥95%
3-Cyano-4,6-dimethyl-2-pyridine carboxylic acid

Hazard: Irritant

Formula: C₉H₈N₂O₂
MW: 176.18 g/mol
Melting Pt: 144 °C (dec.)
MDL Number: MFCD14584826
CAS Number: 1221791-59-8

Order Now

Specification Test Results

Melting point 144°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR