1-Thia-4-azaspiro[4.4]nonan-3-one ≥95%

Supplier: Matrix Scientific

054535-500MG 054535-1G
101907-672EA 278.46 USD
101907-672 101907-674
1-Thia-4-azaspiro[4.4]nonan-3-one ≥95%
1-Thia-4-azaspiro[4.4]nonan-3-one

Hazard: Irritant

Formula: C₇H₁₁NOS
Melting Pt: 163…165 °C
MDL Number: MFCD00101404
CAS Number: 72752-82-0

Order Now

Specification Test Results

Melting point 163-165°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR