Ethyl-3-cyano-4,6-dimethyl-2-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

054533-500MG 054533-1G 054533-5G
101907-662EA 375.38 USD
101907-662 101907-664 101907-666
Ethyl-3-cyano-4,6-dimethyl-2-pyridinecarboxylate ≥95%
Ethyl-3-cyano-4,6-dimethyl-2-pyridinecarboxylate

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₂
MW: 204.23 g/mol
Melting Pt: 58…60 °C
MDL Number: MFCD14584824
CAS Number: 1221792-05-7

Order Now

Specification Test Results

Melting point 58-60°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR