N1-(2-Aminobenzyl)-1,2-benzenediamine ≥95%

Supplier: Matrix Scientific

054531-500MG 054531-1G
101907-656EA 556.92 USD
101907-656 101907-658
N1-(2-Aminobenzyl)-1,2-benzenediamine ≥95%
N1-(2-Aminobenzyl)-1,2-benzenediamine

Hazard: Irritant

Formula: C₁₃H₁₅N₃
Melting Pt: 98…100 °C
MDL Number: MFCD14584822
CAS Number: 14573-33-2

Order Now

Specification Test Results

Melting point 98-100°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR